missing translation for 'onlineSavingsMsg'
Learn More
Thermo Scientific™ Nalgene™ Wide-Mouth LDPE Wash Bottles Product Code.: 10152591
Add to basket

Thermo Scientific™ Nalgene™ Wide-Mouth LDPE Wash Bottles
GREENER_CHOICE

Product Code. 10152591
500 mL, 53 mm
Click to view available options
Capacity (Metric):
500 mL
1000 mL
Closure Size:
53 mm
63 mm
This item is not returnable. View return policy

Product Code. 10152591

Brand: Thermo Scientific™ 24070500

Edge
Add to basket
Add to basket
This item is not returnable. View return policy

Pour without sputter with these wide-mouth LDPE wash bottles which feature through-the-top closure and stem molded in one piece.

High quality squeeze bottles with wide mouth for easy filling. Thermo Scientific™ Nalgene™ Wide Mouth LDPE Wash Bottles are made from lab quality low density polyethylene (LDPE) for dependably low leachables and chemical compatability with a wide range of lab reagents. Enjoy smooth, precision, sputter-free fluid dispensing with an ergonomic soft squeeze. Ideal for rinsing tubes and lab apparatus, bringing volumetric flasks to final volume and other precision rinsing and solution dispensing needs. The dispensing nozzle is molded in one piece with the bottle closure eliminating risk of separation when the bottle is squeezed. Cut back tip to increase flow.

  • Leakproof† bottle comes with polypropylene copolymer draw tube and polypropylene screw closure.
  • Tip can be cut back for increased flow.
  • Tight fit of draw tube and closure prevents separation when squeezing bottle.
Greener Choice Claims
  • Environmental benefits include:
    • Renewable Energy
TRUSTED_SUSTAINABILITY

Specifications

Capacity 16 oz., 500 mL
Capacity (Metric) 500 mL
Capacity (English) 16 oz.
Color Translucent
Description Polypropylene copolymer draw tube
Material LDPE
No. per Case 24
No. per Pack 4
Closure Included Yes
Graduated No
Mouth Wide
Shape Cylindrical
Height (Metric) 216 mm
Product Type Wash Bottle
Height (English) 8.5 in.
Closure Material Polypropylene
Closure Size 53 mm
Diameter (Metric) 73.2 mm
Closure Type Screw Cap
Through the Closure Dispensing Yes
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number1408586 Certificate TypeCertificate of Compliance Date21/06/2024 Catalog Number2407-0500
Lot Number1403111 Certificate TypeCertificate of Compliance Date26/04/2024 Catalog Number2407-0500
Lot Number1396022 Certificate TypeCertificate of Compliance Date15/12/2023 Catalog Number2407-0500
Lot Number1394321 Certificate TypeCertificate of Compliance Date26/10/2023 Catalog Number2407-0500
Lot Number1391626 Certificate TypeCertificate of Compliance Date24/09/2023 Catalog Number2407-0500
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ Nalgene™ Wide-Mouth LDPE Wash Bottles >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.