missing translation for 'onlineSavingsMsg'
Learn More

Naloxone hydrochloride, Tocris Bioscience™

Product Code. 30001105
missing translation for 'orderingAttributeHoverText'
Quantity:
100 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Non-selective opioid antagonist

Opioid antagonist.

Specifications

CAS 357-08-4
Molecular Formula C19H22ClNO4
Quantity 100 mg
Synonym naloxone hydrochloride, naloxone hcl, narcan, evzio, unii-f850569pqr, nalonee, 17-allyl-4,5alpha-epoxy-3,14-dihydroxymorphinan-6-one hydrochloride, narcan tn, naloxonehydrochloride
InChI Key RGPDIGOSVORSAK-STHHAXOLSA-N
SMILES C=CCN1CCC23C4C(=O)CCC2(C1CC5=C3C(=C(C=C5)O)O4)O.Cl
IUPAC Name (4R,4aS,7aR,12bS)-4a,9-dihydroxy-3-prop-2-enyl-2,4,5,6,7a,13-hexahydro-1H-4,12-methanobenzofuro[3,2-e]isoquinoline-7-one;hydrochloride
Molecular Weight (g/mol) 363.838
PubChem CID 5464092
Formula Weight 363.84
Percent Purity >99%
Chemical Name or Material Naloxone hydrochloride
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'