missing translation for 'onlineSavingsMsg'
Learn More

Nalpha-Acetyl-D-arginine dihydrate, 98%, Thermo Scientific Chemicals

Product Code. 11335209
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 210545-23-6
Molecular Formula C8H16N4O3
Molecular Weight (g/mol) 216.24
MDL Number MFCD00209627
InChI Key SNEIUMQYRCDYCH-ZCFIWIBFSA-N
Synonym ac-d-arg-oh, nalpha-acetyl-d-arginine dihydrate, n-alpha-acetyl-d-arginine dihydrate, 2r-5-carbamimidamido-2-acetamidopentanoic acid dihydrate, n-?-acetyl-d-arginine dihydrate, n-, a-acetyl-d-arginine dihydrate, 2r-5-diaminomethylidene amino-2-acetamidopentanoic acid dihydrate
PubChem CID 72698573
IUPAC Name (2R)-5-{[amino(iminiumyl)methyl]amino}-2-acetamidopentanoate
SMILES CC(=O)N[C@H](CCCNC(N)=[NH2+])C([O-])=O

Specifications

Melting Point ∼270°C
Formula Weight 252.29 (216.25 Anhydrous)
Percent Purity 99%
Quantity 1 g
Chemical Name or Material Nalpha-Acetyl-D-arginine dihydrate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'