missing translation for 'onlineSavingsMsg'
Learn More
Nalpha-Boc-D-asparagine, 95%, Thermo Scientific Chemicals Product Code.: 15407628

Nalpha-Boc-D-asparagine, 95%, Thermo Scientific Chemicals

Product Code. 15407628
5 g
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy

Product Code. 15407628

Brand: Thermo Scientific Alfa Aesar H61121.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 75647-01-7
Molecular Formula C9H16N2O5
Molecular Weight (g/mol) 232.236
MDL Number MFCD00065558
InChI Key FYYSQDHBALBGHX-RXMQYKEDSA-N
Synonym boc-d-asparagine, boc-d-asn-oh, n-alpha-boc-d-asparagine, boc-l-asn-oh, nalpha-tert-butoxycarbonyl-d-asparagine, nalpha-boc-d-asparagine, n-boc-d-asparagine, n-alpha-t-butyloxycarbonyl-d-asparagine, d-asparagine, n2-1,1-dimethylethoxy carbonyl, 2r-2-tert-butoxycarbonyl amino-3-carbamoylpropanoic acid
PubChem CID 1623168
IUPAC Name (2R)-4-amino-2-[(2-methylpropan-2-yl)oxycarbonylamino]-4-oxobutanoic acid
SMILES CC(C)(C)OC(=O)NC(CC(=O)N)C(=O)O

Specifications

Color White
Beilstein 4843040
Quantity 5 g
Formula Weight 232.23
Percent Purity 95%
Physical Form Powder
Chemical Name or Material Nalpha-Boc-D-asparagine

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Nalpha-Boc-D-asparagine, 95%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.