Learn More
Nalpha-Boc-L-lysine, 97%, Thermo Scientific Chemicals
102.51 € - 841.19 €
Chemical Identifiers
CAS | 13734-28-6 |
---|---|
Molecular Formula | C11H22N2O4 |
Molecular Weight (g/mol) | 246.31 |
MDL Number | MFCD00038203 |
InChI Key | DQUHYEDEGRNAFO-SVGMAFHSNA-N |
Synonym | boc-lys-oh, boc-lysine, n-boc-l-lysine, boc-l-lysine, s-6-amino-2-tert-butoxycarbonyl amino hexanoic acid, nalpha-tert-butoxycarbonyl-l-lysine, n-alpha-tert-butoxycarbonyl-l-lysine, nalpha-boc-l-lysine, boc-lys, n alpha-boc-l-lysine |
PubChem CID | 2733284 |
IUPAC Name | (2S)-6-amino-2-[(2-methylpropan-2-yl)oxycarbonylamino]hexanoic acid |
SMILES | CC(C)(C)OC(=O)N[C@@H](CCCCN)C(O)=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15480789
|
Thermo Scientific Alfa Aesar
H63859.06 |
5 g |
102.51 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15490789
|
Thermo Scientific Alfa Aesar
H63859.14 |
25 g |
314.57 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15400799
|
Thermo Scientific Alfa Aesar
H63859.22 |
100 g |
841.19 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
N-Boc-L-lysine is a protected analogue of a major amino acid participating in the binding of AcH to proteins. N-Boc-L-lysine as well as other t-butoxycarbonyl (Boc) amino acids can be converted to their respective amino acids via cleavage of the urethane bond.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsN-Boc-L-lysine is a protected analogue of a major amino acid participating in the binding of AcH to proteins. N-Boc-L-lysine as well as other t-butoxycarbonyl (Boc) amino acids can be converted to their respective amino acids via cleavage of the urethane bond.
Solubility
Soluble in water. Slightly soluble in methanol.
Notes
Refrigerate. Keep container tightly closed. Store away from oxidizing agent.
Chemical Identifiers
13734-28-6 | |
246.31 | |
DQUHYEDEGRNAFO-SVGMAFHSNA-N | |
2733284 | |
CC(C)(C)OC(=O)N[C@@H](CCCCN)C(O)=O |
C11H22N2O4 | |
MFCD00038203 | |
boc-lys-oh, boc-lysine, n-boc-l-lysine, boc-l-lysine, s-6-amino-2-tert-butoxycarbonyl amino hexanoic acid, nalpha-tert-butoxycarbonyl-l-lysine, n-alpha-tert-butoxycarbonyl-l-lysine, nalpha-boc-l-lysine, boc-lys, n alpha-boc-l-lysine | |
(2S)-6-amino-2-[(2-methylpropan-2-yl)oxycarbonylamino]hexanoic acid |
Specifications
∼205°C (decomposition) | |
White | |
MFCD00038203 | |
boc-lys-oh, boc-lysine, n-boc-l-lysine, boc-l-lysine, s-6-amino-2-tert-butoxycarbonyl amino hexanoic acid, nalpha-tert-butoxycarbonyl-l-lysine, n-alpha-tert-butoxycarbonyl-l-lysine, nalpha-boc-l-lysine, boc-lys, n alpha-boc-l-lysine | |
DQUHYEDEGRNAFO-SVGMAFHSNA-N | |
(2S)-6-amino-2-[(2-methylpropan-2-yl)oxycarbonylamino]hexanoic acid | |
2733284 | |
97% | |
Nalpha-Boc-L-lysine |
13734-28-6 | |
C11H22N2O4 | |
4252546 | |
Soluble in water. Slightly soluble in methanol. | |
CC(C)(C)OC(=O)N[C@@H](CCCCN)C(O)=O | |
246.31 | |
246.3 | |
Powder |
Safety and Handling
missing translation for 'einecsNumber' : 237-303-4
missing translation for 'tsca' : No
missing translation for 'storageNote' : 2° to 8°C
Recommended Storage : Keep cold
RUO – Research Use Only