missing translation for 'onlineSavingsMsg'
Learn More

Nano-Secondary™ Alpaca, anti-Human, CoraLite Plus 488, Chromotek

Alpaca Recombinant Monoclonal Antibody

110.77 € - 390.15 €

Specifications

Antigen Nano-Secondary™
Conjugate CoraLite Plus 488
Form Liquid
Host Species Alpaca
Target Species Human
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18018945
View Documents
Chromotek
shuGCL488-2-10
10 μL
110.77 €
10µL
Please to purchase this item. Need a web account? Register with us today!
18015475
View Documents
Chromotek
shuGCL488-2-100
100 μL
390.15 €
100µL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Nano-Secondary™ alpaca anti-human IgG, recombinant VHH, is a human IgG-specific (IgG1, IgG2, IgG3, IgG4) antibody. This secondary antibody product consists of a Nanobody that binds to the Fc-domain of human IgG with high affinity and specificity.

Specifications

Specifications

Nano-Secondary™
Liquid
Human
Secondary
CoraLite Plus 488
Alpaca
IgG
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.