Learn More
Nano-Secondary™ Alpaca, anti-Human, CoraLite Plus 555, Chromotek
Alpaca Recombinant Monoclonal Antibody
Brand: Chromotek shuGCL555-2-100
This item is not returnable.
View return policy
Description
Nano-Secondary™ alpaca anti-human IgG, recombinant VHH, is a human IgG-specific (IgG1, IgG2, IgG3, IgG4) antibody. This secondary antibody product consists of a Nanobody that binds to the Fc-domain of human IgG with high affinity and specificity.
Specifications
Nano-Secondary™ | |
Alpaca | |
Secondary | |
Liquid |
CoraLite Plus 555 | |
100 μL | |
Human | |
IgG |