missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ NanoDrop Lite Plus Spectrophotometers

Quickly and easily assess sample concentration and purity ratios with the entry-level Thermo Scientific™ NanoDrop™ Lite Plus Microvolume Spectrophotometer. The instrument delivers accurate measurements in an easy-to-use, affordable package. Ideal for teaching and busy research labs.

Specifications

Absorbance Range 0.04 to 30 Abs (10 mm equivalent)
Certifications/Compliance UL/CSA and CE
Compatibility Local Control Only
Connections USB 2.0 Port
Depth (Metric) 22 cm
View More Specs

Description

Description

Quickly and easily assess sample concentration and purity ratios with the Thermo Scientific™ NanoDrop™ Lite Plus Microvolume Spectrophotometer, the entry-level complement to the full-featured Thermo Scientific™ NanoDrop™ One/OneC and NanoDrop Eight Spectrophotometers. While offering fewer features than our premium models, this instrument delivers fast, accurate measurements in an easy-to-use, affordable package.
 
The NanoDrop Lite Plus Spectrophotometer is a valuable teaching tool and is ideal for quality check measurements in busy research labs as well. Measure purified DNA, RNA, and protein concentration up to 30 Abs and calculate critical A260/A280 and A260/A230 purity ratios using trusted NanoDrop technology.
 

The NanoDrop Lite Plus Spectrophotometer is designed for routine measurements of nucleic acid and protein samples using only 1–2 μL. Our pioneering sample-retention technology eliminates the need to dilute highly concentrated samples, making measurements easy. Simply pipette your sample directly onto the optical measurement pedestal, measure, and wipe clean with a lint-free wipe. Its compact design, with built-in controls and software, means the NanoDrop Lite Plus Spectrophotometer takes up little space on your benchtop.
 
With an intuitive user interface and modern industrial design, the NanoDrop Lite Plus Spectrophotometer is the instrument of choice for basic microvolume measurements. Even if budgets are tight, your lab can have a NanoDrop, too.
 
Confidence in your results
  • Standard measurements
    • Measures purified DNA, RNA, and protein concentration up to 30 Abs
      • Up to 1,500 ng/µL dsDNA
      • Up to 1,200 ng/µL RNA
      • Up to 45 mg/mL BS
    • Calculates critical A260/A280 and A260/A230 purity ratios
  • Fast, accurate measurements from only 1–2 µL—saves precious samples, no waste
    • Sample-retention technology uses surface tension to hold the sample in place—no cuvettes or expensive consumables are needed
  • Intuitive interface
    • Built-in touchscreen operation
    • No computer required
  • Data storage and transfer
    • Last 1,000 measurements saved locally; export data to a USB device
  • Optional printer available for freezer-compatible labels (not included)
  • Compact and portable—fits on any lab bench, ready for your next experiment
  • Affordably priced—stretch tight budgets
Reach your goals
  • Academic teaching—educate students on molecular biology principles and experimental techniques on an industry-leading instrument
  • Academic research labs—facilitate your research and publication of your novel research findings
  • Biotech start-ups—accelerate the develop of new products
 
Specifications

Specifications

0.04 to 30 Abs (10 mm equivalent)
Local Control Only
22 cm
LCD Touchscreen
Microvolume UV-Vis Spectrophotometer
Nucleic Acid Quantification, DNA Quantification, RNA Quantification, Protein Quantification
Stainless Steel (303), Quartz Fiber
1.0 mm and 0.2 mm (auto adjust)
Standalone control with data export via USB Flash Drive
27 cm
UL/CSA and CE
USB 2.0 Port
2048-element CMOS Linear Image Sensor
Xenon Flash
230 nm, 260 nm, 280 nm
22 cm
1
≤ 1.8 nm (FWHM at Hg 254 nm)
One year
Videos
SDS
Documents

Documents

Product Certifications