Learn More
Thermo Scientific™ NanoDrop™ Lite Plus Printer Accessory
Optional printers for use with your Thermo Scientific™ NanoDrop™ Lite Plus or NanoDrop Lite Spectrophotometer
Brand: Thermo Scientific™ NDLP-PRNTR-MOD
Additional Details : Weight : 1.60000kg
Description
Use an optional printer with your NanoDrop™ Lite Plus Spectrophotometer. Print data on freezer-compatible labels.
Print sample data on a cryogenic label
- Sample number
- Date
- Time of Measurement
- Sample Name
- Assay
- Concentration
- Purity Ratio
For use with the NanoDrop Lite Spectrophotometer
Specifications
Printer Accessory for NanoDrop Lite Plus | |
NanoDrop Lite Plus Spectrophotometer |