Learn More
Implen™ NanoPhotometer™ C40 Cuvette UV-VIS Spectrophotometer
For cuvette applications. Suitable for all common photometric measurements like protein assays, kinetics and standard curve. Intuitive touchscreen operation, small footprint and network integration for convenient lab bench operation. Recalibration-free patented technology.
7650.00 € - 8450.00 €
Specifications
Absorbance Accuracy | < 1.75 % at 0.7 A (0.67 mm path) at 280 nm of the reading |
---|---|
Absorbance Range | 0 to 2.6 A |
Beam Center Height | 8.5 mm |
Connections | 2x USB A, USB B, HDMI, Ethernet, WiFi |
Detector Type | 3648-element CCD Array |
Product Code | Brand | Includes | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Includes | Price | Quantity & Availability | |||||
15528783
|
Implen™
C40-Mobile |
Battery, Power Supply, USB Cable, Dust Cover, Software and Manual |
8450.00 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15598783
|
Implen™
C40-Touch |
Power Supply, USB Cable, Dust Cover, Software and Manual |
7650.00 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The Implen NanoPhotometer™ C40 UV/VIS spectrophotometer offers full scan data from 200 - 900 nm for all cuvette based photometric applications in Life Sciences. It can be upgraded to NanoVolume using the Implen Submicoliter Cell. The stand-alone design with an integrated touchscreen and small footprint make it ideal for lab bench operation.
No recalibration is necessary over the entire lifetime. Optional CFR21 software available for 21 CFR part 11 compliance for GxP environment.
- Detection range dsDNA 0.1 ng/μL - 130 ng/μL
- Detection range BSA 0.003 mg/mL - 3.7 mg/mL
- Photometric Range 0 - 2.6 A
- Scan range 200 - 900 nm
- Center Height (Z-Height) 8.5 mm
- Suitable for all cuvettes with outside dimensions 12.5 x 12.5 mm
- Temperature controlled cuvette port 37°C
- Measure time for full scan range: 2.5 seconds
- Preprogrammed applications for (labeled) nucleic acids (DNA, RNA, oligos), (labeled) proteins, protein assays (Bradford, BCA, Lowry), kinetics, OD600 and manual methods
- Recalibration-free due to True Path Technology™
- Sample ControlTM and Blank ControlTM for analysis of sample quality and identification of contaminants
- Control options: Onboard with built-in 7” Touchscreen, Computer and Tablet
- In and Output Ports 2x USB A, USB B, HDMI, Ethernet, Wi-Fi
- 21 CFR part 11 compliant software (optional)
- Built-in file server
- LIMS integration through REST API
- Built-in battery pack, operation time: up to 8 h
- Made in Germany
Specifications
< 1.75 % at 0.7 A (0.67 mm path) at 280 nm of the reading | |
8.5 mm | |
3648-element CCD Array | |
200 x 200 x 120 mm | |
Xenon Flash | |
Spectrophotometer | |
200 to 900 nm | |
2.5 sec | |
37 °C ± 0.5 °C | |
Linux based NPOS | |
± 0.75 nm |
0 to 2.6 A | |
2x USB A, USB B, HDMI, Ethernet, WiFi | |
7 in. Glove Compatible Touchscreen, 1024 x 600 pixels | |
English, French, German, Spanish, Portuguese, Russian, Chinese, Japanese | |
<1.8 nm | |
90/250 V | |
Nucleic Acids, Protein UV, Protein Assays, Kinetics, OD600, Wavelength, Wavescan, Standard Curve, Absorbance Ratio, Concentration, Custom | |
1 | |
10, 5, 2, 1, 0.5 mm | |
Windows 7, 8, 10 (32 & 64 bit); OS X, iOS; Android OS |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.