Learn More
Implen™ NanoPhotometer™ IQ/OQ Documentation and Standard Solution
In compliance with international standards in regulated environments, documented verification is provided with Installation Qualification and Operation Qualification (IQ/OQ) Package Conforming Data to verify that instruments are working according to specifications in case of an audit.
1290.00 €
Specifications
Certifications/Compliance | According to Internal SOP |
---|---|
Calibration Type | Self-calibration |
Product Type | IQ/OQ Package Conforming Data |
Product Line | NanoPhotometer |
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | |||||
15533214
|
Implen™
N-60-Q |
NanoPhotometer N60 |
1290.00 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16658323
|
Implen™
N-120-Q |
NanoPhotometer N120 |
1290.00 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- Documentation provided on a USB flash drive as an Excel file
- Includes NanoPhotometer™ appropriate standard solution (N-568-S or N-120-S)
Specifications
According to Internal SOP | |
Self-calibration | |
IQ/OQ Package Conforming Data | |
NanoPhotometer |
For Research Use Only