missing translation for 'onlineSavingsMsg'
Learn More

Implen™ NanoPhotometer™ N120 Multichannel Micro-Volume UV-VIS Spectrophotometer

The Implen NanoPhotometer™ N120 scans up to 12 samples in just 20 seconds. Quantifying DNA, RNA and proteins has never been faster. Intuitive touchscreen operation, small footprint and network integration for convenient lab bench operation. Recalibration-free patented technology Made in Germany.

Brand:  Implen™ N120-Touch

 View more versions of this product

Product Code. 16309021

  • 18850.00 € / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

The NanoPhotometer™ N120 is the latest member of the NanoPhotometer™ family. It provides high-throughput measurements of up to 12 samples without consumables, e.g. DNA or proteins, at once. The stand-alone design with an integrated touchscreen and small footprint make it ideal for lab bench operation.

Disturbing surface tension and evaporation effects are eliminated due to the compression of the sample. No recalibration is necessary over the entire lifetime. Optional CFR21 software available for 21 CFR part 11 compliance for GxP environment.

  • Sample number 1 - 12
  • Detection range dsDNA 2 ng/μL - 8,000 ng/μL
  • Detection range BSA 0.06 mg/mL - 230 mg/mL
  • Minimum sample Size 2.0 μL
  • Scan range 200 - 900 nm
  • Measure time for full scan range: 1.7 - 2.5 seconds
  • Preprogrammed applications for (labeled) nucleic acids (DNA, RNA, oligos), (labeled) proteins, protein assays (Bradford, BCA, Lowry), kinetics, OD600 and manual methods
  • Auto Sample function starts automatically once sample arm is closed
  • Recalibration-free due to True Path Technology™
  • Independent from surface tension of sample due to Sample Compression Technology™
  • Sample ControlTM and Blank ControlTM for analysis of sample quality and identification of contaminants
  • Control options: Onboard with built-in 7” Touchscreen, Computer and Tablet
  • In & Output Ports 2x USB A, USB B, Ethernet, Wi-Fi
  • 21 CFR part 11 compliant software (optional)
  • Built-in file server
  • LIMS integration through REST API
  • Made in Germany
Specifications

Specifications

< 1.75% at 0.7 A at 280 nm of the reading
3648-element CCD Array
20 x 20 x 12 cm
Xenon Flash
90/250 V
Nucleic Acids, Protein UV, Protein Assays, Kinetics, OD600, Wavelength, Wavescan, Standard Curve, Absorbance Ratio, Concentration, Custom
N120-Touch
1 and 0.125 mm
2 to 3.5 μL
Windows 7, 8, 10 (32 & 64 bit); OS X, iOS; Android OS
5 kg
0.04 to 160 A
7 in. Glove Compatible Touchscreen, 1024 x 600 pixels
English, French, German, Spanish, Portuguese, Russian, Chinese, Japanese
<2.5 nm
200 to 900 nm
1.7 - 2.5 seconds
1 to 12
Linux based NPOS
< 1% @ 240 nm using NaI
± 0.75 nm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
View accessory products
Special Offers

Special Offers

For Research Use Only