Learn More
Cytiva Nanosep™ Centrifugal Devices with Omega™ Membrane
Use Nanosep™ devices to concentrate, purify, and desalt peptides, proteins, oligonucleotides, DNA, and RNA.
Brand: Cytiva OD030C34
534.01 EUR valid until 2024-12-31
Use promo code "21930" to get your promotional price.
This item is not returnable.
View return policy
Description
- Simple, reliable,concentrating and desalting of 50 to 500 μL samples
- This product is not approved for use in medical, clinical, surgical or other patient applications
Specifications
Red | |
PES | |
Non-sterile | |
100/Pk. |
EFA: 0.3 cm2 | |
30 kDa | |
Polypropylene | |
50 to 500 μL (Sample) |