Learn More
Cytiva Nanosep™ Centrifugal Devices with Omega™ Membrane
Use Nanosep™ devices to concentrate, purify, and desalt peptides, proteins, oligonucleotides, DNA, and RNA.
Brand:  Cytiva OD003C33
135.85 EUR valid until 2024-12-31
Use promo code "21930" to get your promotional price.
 View more versions of this product
Product Code. 17194771
  • 143.00 € / Pack of 24
Estimated Shipment: 10-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Simple, reliable,concentrating and desalting of 50 to 500 μL samples
  • This product is not approved for use in medical, clinical, surgical or other patient applications
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Gray
PES
Non-sterile
24/Pk.
EFA: 0.3 cm2
3 kDa
Polypropylene
50 to 500 μL (Sample)