missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Nanosep™ MF Centrifugal Devices with Bio-Inert™ Membrane

Use Nanosep™ devices to concentrate, purify, and desalt peptides, proteins, oligonucleotides, DNA, and RNA.

Brand:  Cytiva ODM45C34

427.49 EUR valid until 2025-12-31
Use promo code "24320" to get your promotional price.


 View more versions of this product

Product Code. 17134801

  • 530.00 € / Pack of 100
In Stock
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Simple, reliable,concentrating and desalting of 50 to 500 μL samples
  • This product is not approved for use in medical, clinical, surgical or other patient applications
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Wildberry
Bio-Inert (modified nylon)
Non-sterile
100/Pk.
EFA: 0.3 cm2
0.45 μm
Polypropylene
50 to 500 μL (Sample)
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

One moment while we fetch your results.
Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Cytiva Nanosep™ MF Centrifugal Devices with Bio-Inert™ Membrane > 0.45 μm; 100/Pk.

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.