missing translation for 'onlineSavingsMsg'
Learn More

Cytiva (formerly Pall Lab) Nanosep™ MF Centrifugal Devices with Bio-Inert™ Membrane

Use Nanosep™ devices to concentrate, purify, and desalt peptides, proteins, oligonucleotides, DNA, and RNA.

Brand:  Cytiva (formerly Pall Lab) ODM02C33

134.01 EUR valid until 2024-12-31
Use promo code "21930" to get your promotional price.


 View more versions of this product

Product Code. 17194791

  • 143.00 € / Pack of 24
Estimated Shipment: 03-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Simple, reliable,concentrating and desalting of 50 to 500 μL samples
  • This product is not approved for use in medical, clinical, surgical or other patient applications
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Aqua
Bio-Inert (modified nylon)
Non-sterile
24/Pk.
EFA: 0.3 cm2
0.2 μm
Polypropylene
50 to 500 μL (Sample)
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers