missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Naphthalene, 99+%, scintillation grade, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
250 g
1 kg
Packaging:
Glass Bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For fluorescence/luminescence spectroscopy
Specifications
Specifications
Boiling Point | 218.0°C |
Melting Point | 79.0°C to 82.0°C |
Physical Form | Powder |
CAS Min % | 99.0 |
CAS Max % | 100.0 |
Density | 0.9900g/mL |
Color | Beige to Red/Salmon |
Quantity | 250 g |
Flash Point | 78°C |
Assay Percent Range | 99+% |
Show More |
Safety and Handling
Product Identifier
- Naphthalene
- Warning
- Flammable solid Category 2
- Acute toxicity Category 4
- Carcinogenicity Category 2
- ACUTE AQUATIC Acute 1
- LONG-TERM AQUATIC HAZARD Chronic 1
- H228-Flammable solid.
- H302-Harmful if swallowed.
- H351-Suspected of causing cancer.
- H410-Very toxic to aquatic life with long lasting effects.
- P201-Obtain special instructions before use.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P264-Wash thoroughly after handling.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- P273-Avoid release to the environment.
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction