missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Naphthalene-d8, for NMR, 98+ atom % D
Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass Bottle
Unit Size:
1g
5g
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For NMR
Specifications
Specifications
| Physical Form | Crystals or Crystalline Powder |
| Color | White |
| Deuterium (D) | 98.0% min. |
| Flash Point | 78°C |
| Packaging | Glass Bottle |
| Quantity | 1 g |
| Melting Point | 81.0°C to 83.0°C |
| Linear Formula | C10D8 |
| Formula Weight | 136.22 |
| Grade | NMR |
| Show More |
Safety and Handling
Product Identifier
- Naphthalene-d8
- Warning
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 1
- Carcinogenicity Category 2
- Flammable solid Category 2
- H228-Flammable solid.
- H302-Harmful if swallowed.
- H351-Suspected of causing cancer.
- H410-Very toxic to aquatic life with long lasting effects.
- P201-Obtain special instructions before use.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P264-Wash thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P370+P378-In case of fire: Use to extinguish.
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction