missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nasco Meat and Poultry Carcass Sampling Kit (18 oz.) with PE Gloves and Buffer
Designed for sampling meat and turkey carcasses to test for the presence of E. coli
Brand: Nasco B01324WA
This item is not returnable.
View return policy
Description
- One pair of non-handed (Size: L), sterile polyethylene gloves
- 25 mL buffered peptone water (expiration date marked on the box)
- Whirl-Pak™ Speci-Sponge™ bag
- Two disposable sterile templates, one for meat carcasses (10 x 10 cm) and one for turkeys (5 x 10 cm)
- Disposable template eliminates the need for re-sanitizing reusable templates, saving time and money and making it easier to transport the template to the carcass
- Templates form to the contours of the carcass, making it easier to swab within the required area
- Gloves and templates are packaged together in a sealed plastic bag that has a perforation for easy opening
Order Info
Box of 25 kits
Specifications
18 oz. | |
Meat and Poultry Carcass Sampling | |
25 Kits/Box |
532 mL | |
One pair of non-handed (Size: L), sterile polyethylene gloves; 25 mL buffered peptone water (expiration date marked on the box); Whirl-Pak™ Speci-Sponge™ bag; Two disposable sterile templates, one for meat carcasses (10 x 10 cm) and one for turkeys (5 x 10 cm) |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction