Learn More
Nasco Whirl-Pak™ Stand-Up Sample Bags
GREENER_CHOICE
Stand-Up Bags make it very easy to add material to the bag and to remove sample for testing or analysis. The flat bottom ensures stability and the wire tabs hold the top open for easy access.
Brand:  Nasco B01365WA
 View more versions of this product
Product Code. 11726873
  • 179.00 € / Pack of 500
In Stock
to see stock.
Edge
Explore more special offers
Add to basket
Description
Description
White, write-on strip makes sample identification easier and more convenient
  • Paper-like finish virtually eliminates ink smearing
  • Surface that will accept a variety of writing instruments
  • Sterilized after manufacturing
Specifications
Specifications
Whirl-Pak™ Stand-Up Bag
532 mL
0.076 mm
82°C
23 x 11.5 cm (9 x 4.5 in.)
23 cm
11.5 cm
18 oz.
Sterile
500/Pk.
3 mil
9 in.
4.5 in.
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
View accessory products