Learn More
Thermo Scientific™ National Assembled Standard Opening Vial, Cap and Septum Kits
Thermo Scientific™ National Assembled Standard Opening Vial, Cap and Septum Kits provide convenience, efficiency and stackable storage options.
Brand:  Thermo Scientific™ C4013-019A
 View more versions of this product
Product Code. 11941613
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
Thermo Scientific™ National Assembled Standard Opening Vial, Cap and Septum Kits provide convenience, efficiency and stackable storage options.
  • Packaged in convenient vial trays with clear covers or in economical polybags
  • Vial trays are stackable for easy storage
  • All caps are manufactured from virgin polypropylene unless noted
Includes: 
100 vials with pre-attached caps and septa
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Black
Amber
2 mL
Polypropylene
8-425
Amber Gkass; PP Black Cap; White PTFE septum; in polybag
PTFE
White PTFE
Polypropylene