Learn More
Thermo Scientific™ National Assembled Standard Opening Vial, Cap and Septum Kits
Thermo Scientific™ National Assembled Standard Opening Vial, Cap and Septum Kits provide convenience, efficiency and stackable storage options.
Brand: Thermo Scientific™ C4013-486A
This item is not returnable.
View return policy
Description
Thermo Scientific™ National Assembled Standard Opening Vial, Cap and Septum Kits provide convenience, efficiency and stackable storage options.
- Packaged in convenient vial trays with clear covers or in economical polybags
- Vial trays are stackable for easy storage
- All caps are manufactured from virgin polypropylene unless noted
Includes:
100 vials with pre-attached caps and septa
Specifications
Black | |
Clear | |
2 mL | |
Phenolic | |
8-425 |
Clear Glass; Phenolic Black Cap; PTFE/Silicone/PTFE septum; in storage tray | |
PTFE, Silicone | |
PTFE/Silicone/PTFE | |
Phenolic |