missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Components for Vial Convenience Kits
Vials, caps and septa for use with Thermo Scientific™ National Vial Convenience Kits.
Brand: Thermo Scientific™ B7920-VO
This item is not returnable.
View return policy
Description
Vials, caps and septa for use with Thermo Scientific™ National vial convenience kits.

Specifications
Clear 20mL vials, 28 x 57mm | |
28 mm | |
National | |
Screw Cap | |
100 Pk. | |
Vial |
Clear | |
20 mL | |
20 mL | |
57 mm | |
Certificate of Analysis (COA), Certificate of Conformance (COC) | |
Thermo Scientific™ National Vial Convenience Kits |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction