missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Components for Vial Convenience Kits

Vials, caps and septa for use with Thermo Scientific™ National Vial Convenience Kits.

Brand:  Thermo Scientific™ B7921-VO

 View more versions of this product

Product Code. 12438646

  • 51.14 € / Pack of 100
In Stock
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Vials, caps and septa for use with Thermo Scientific™ National vial convenience kits.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Amber 20mL vials, 28 x 57mm
28 mm
National
Screw Cap
100 Pk.
Vial
Amber
20 mL
20 mL
57 mm
Certificate of Analysis (COA), Certificate of Conformance (COC)
Thermo Scientific™ National Vial Convenience Kits
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.