Learn More
Memmert™ Natural Convection Hot Air Sterilizer with SingleDISPLAY
Sterilize a variety of containers and instruments with this hot air sterilizer. Memmert™ Natural Convection Hot Air Sterilizers with SingleDISPLAY allow adjustment of pre-heated fresh air admixture by air flap control in 10% steps. Safer than disinfection.
1320.00 € - 6705.00 €
Specifications
For Use With (Application) | Applied for sterilisation of medical materials, complete killing of highly resistant microorganisms, special application of depyrogenisation |
---|
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
12895843
|
Memmert™
SN110 |
108 L |
2160.00 €
Each |
Estimated Shipment: 17-12-2024
Log in to see stock. |
|||||
12805853
|
Memmert™
SN160 |
161 L |
2870.00 €
Each |
Estimated Shipment: 28-01-2025
Log in to see stock. |
|||||
12815853
|
Memmert™
SN260 |
256 L |
3800.00 €
Each |
Estimated Shipment: 28-01-2025
Log in to see stock. |
|||||
12875843
|
Memmert™
SN30 |
32 L |
1320.00 €
Each |
Estimated Shipment: 28-01-2025
Log in to see stock. |
|||||
12825853
|
Memmert™
SN450 |
449 L |
5030.00 €
Each |
Estimated Shipment: 28-01-2025
Log in to see stock. |
|||||
12865843
|
Memmert™
SN55 |
53 L |
1515.00 €
Each |
Estimated Shipment: 28-01-2025
Log in to see stock. |
|||||
12885843
|
Memmert™
SN75 |
74 L |
1865.00 €
Each |
Estimated Shipment: 17-12-2024
Log in to see stock. |
|||||
12835853
|
Memmert™
SN750 |
749 L |
6705.00 €
Each |
Estimated Shipment: 28-01-2025
Log in to see stock. |
|||||
Description
- German, English, Spanish, French, Polish, Czech, Hungarian language settings
- SingleDISPLAY provides adaptive multifunctional digital PID-microprocessor controller with high-definition TFT-color display
- Digital backwards counter with target time setting, adjustable from 1 minute to 99 days
- Process time does not start until the set temperature is reached
- Three freely selectable temperature values
- Adjust temperature (Celsius or Fahrenheit), air flap position, program time, time zones, summertime/wintertime
Specifications
Applied for sterilisation of medical materials, complete killing of highly resistant microorganisms, special application of depyrogenisation |