Learn More
R&L Enterprises™ Semi-Micro Natural Gas Bunsen Burner
An excellent solution for heating, sterilizing and combustion in the laboratory. R&L Enterprises™ Semi-Micro Natural Gas Bunsen Burners are constructed in brass and nickel plated.
Brand:  R&L Enterprises™ 4015/2
Product Code. 12852154
  • 29.65 € / Each
Estimated Shipment: 04-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Constructed using a heavy pressure diecast base painted in a chemically resistant blue acrylic finish
  • Tested for flame quality and leakage prior to packing
Specifications
Specifications
Semi-micro Burner
9 mm
1 Ea.
Natural Gas, Methane
100 mm
0.22 kg