Learn More
Kautex™ LDPE Wide Mouth Bottles Series 303
Designed with a wide opening for easy filling and cleaning. Kautex™ Wide Mouth Bottles are ideal for liquids, pastes and granules.
Brand:  Kautex™ 2000770530
 View more versions of this product
Product Code. 12684940
  • 28.35 € / Pack of 10
Explore more special offers
Description
Description
  • Manufactured from low density polyethylene (LDPE) for flexibility and toughness
  • Natural translucent color, round bottles
  • Easy to fill and clean
  • Ideal for dispensing, sending samples, adhesives, transportation and storage 
  • Includes leakproof screw closure
TRUSTED_SUSTAINABILITY
Specifications
Specifications
200 mL
Round
LDPE
Screw Closure LDPE
No
Wide Mouth Bottle
Dispensing, Sending Samples, Mailing, Transport, Storage
Closure
30.8 mm
1 Ea.
114 mm
Screw Cap