Learn More
Ansell Edmont™ Profil™ Plus 87-850 Series Yellow Natural Rubber Latex Immersion Gloves
Product Code. 12425120
Click to view available options
Unit Size:
12 pairs
Size:
8
9
Product Code. 12425120
Brand: Ansell Edmont™ 87850070
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Medium weight gloves with outstanding grip. Excellent resistance to mechanical hazards and enhanced chemical resistance.100% cotton flocklining makes the glove softer and more comfortable to wear while helping to absorb perspiration. Specially treated to reduce the risk of allergy.
  • Specially treated to reduce the risk of allergy
  • Excellent resistance to mechanical hazards and enhanced chemical resistance
  • 100% cotton flocklining - soft, comfortable and absorbent
  • Ideal for janitorial use and handling in wet conditions
TRUSTED_SUSTAINABILITY
Specifications
Back Material Rubber Latex
Cuff Material Natural Rubber Latex
Coating Material Natural Rubber Latex
Latex Free No
Product Type Immersion Gloves
Style 87-850
Thickness (Metric) 0.55 mm
Glove Cuff Color Yellow
Glove Cuff Style Pinked
Liner Material Cotton Flocking
Quantity 144 Pairs
Resistant To Mechanical hazards and enhanced chemical resistance
Series 87-850