missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BRAND™ Natural Rubber Pipet Filler
Manufactured from 100% natural rubber. BRAND™ Natural Rubber Pipet Filler eliminates mouth pipetting and handling of hazardous material. Available in universal, standard, or flip model, with 2 or 3-valve options.
Brand: BRAND™ 25315
15.19 EUR valid until 2024-11-29
Use promo code "21683" to get your promotional price.
This item is not returnable.
View return policy
Description
- Manufactured from 100% natural rubber.
- Eliminates mouth pipetting and handling of hazardous material.
- Available in universal, standard, or flip model, with 2 or 3-valve options
- 2-valve flip model enables one-handed operation.
Specifications
Valve A: Release air, Valve S: Filling, Valve E: Delivery | |
Universal | |
Filling pipets up to 100mL | |
1 Each |
Natural Rubber | |
100 mL | |
3 valves |