Learn More
Fisherbrand™ Solid Natural Rubber Stoppers
Used for laboratory purposes.
Brand:  Fisherbrand™ RB023/S
 View more versions of this product
Product Code. 10192031
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
  • Made up of natural rubber
  • Red in color
  • Solid stoppers with no holes
Specifications
Specifications
For laboratory use
28 mm
Red
26.5 mm
Stopper
46/55 IRHD
20/Pk.
23 mm
Natural Rubber