Learn More
Abeba™ Nature 4080 Shoes
Leather covered cork footbed is ESD dissipative.
Brand: Abeba™ 4080/43
This item is not returnable.
View return policy
Description
- CE EN ISO 20347:2012 OB A, FO SRC
- Smooth leather
- White
- Slip resistance special rubber outsole sole
- Leather coated cork footbed
- Sanitized treated
- ESD dissipative according to DIN EN 61340
- 2 adjustable instep straps
- velcro fastening
Specifications
CE EN ISO 20347:2012 OB A, FO SRC | |
Cork | |
43 | |
Rubber |
White | |
Leather | |
Slip-resistant | |
4080 |