missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ NAV 2729

Selective ARF6 inhibitor

270.00 € - 1080.00 €

Specifications

Color Yellow
Product Type Inhibitor
CAS 419547-11-8
Molecular Formula C25H17ClN4O3
Formula Weight 456.88g/mol
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18730128
View Documents
Tocris Bioscience™
5986/10
10 mg
270.00 €
10mg
Please to purchase this item. Need a web account? Register with us today!
18755999
View Documents
Tocris Bioscience™
5986/50
50 mg
1080.00 €
50mg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Selective ARF6 inhibitor (IC50 = 1.0 μM). Selective for ARF6 over other ARF family members and small GTPases. Inhibits ARF6 activation in uveal melanoma cells in vitro and blocks downstream signaling pathways of oncogenic GNAQ. Reduces tumorigenesis in mouse model of uveal melanoma.
Specifications

Specifications

Yellow
419547-11-8
456.88g/mol
Store at 4°C
Inhibitor
C25H17ClN4O3
Soluble to 100mM in DMSO
Blocks downstream signaling pathways
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only