missing translation for 'onlineSavingsMsg'
Learn More
NBD chloride, 98%, Thermo Scientific Chemicals Product Code.: 10721451

NBD chloride, 98%, Thermo Scientific Chemicals

Product Code. 10721451
5 g, Glass bottle
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10721451

Brand: Thermo Scientific Acros 172390050

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10199-89-0
Molecular Formula C6H2ClN3O3
Molecular Weight (g/mol) 199.55
MDL Number MFCD00005808
InChI Key IGHBXJSNZCFXNK-UHFFFAOYSA-N
Synonym 4-Chloro-7-nitrobenzofurazan, 4-Chloro-7-nitrobenzo-2-oxa-1, 3-diazole
PubChem CID 25043
ChEBI CHEBI:78878
IUPAC Name 4-chloro-7-nitro-2,1,3-benzoxadiazole
SMILES C1=C(C2=NON=C2C(=C1)Cl)[N+](=O)[O-]

Specifications

Melting Point 96.0°C to 99.0°C
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Solubility Information Solubility in water: soluble.
Infrared Spectrum Authentic
Formula Weight 199.55
Percent Purity 98%
Packaging Glass bottle
Color Brown to Yellow
Quantity 5 g
Physical Form Crystalline Powder
Chemical Name or Material NBD chloride
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0438148 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog Number172390010,172390250,172390050
Lot NumberA0438148 Certificate TypeCertificate of Analysis Date17/01/2024 Catalog Number172390010,172390250,172390050
Lot NumberA0436295 Certificate TypeCertificate of Analysis Date16/11/2023 Catalog Number172390010,172390250,172390050
Lot NumberA0396582 Certificate TypeCertificate of Analysis Date15/12/2022 Catalog Number172390010,172390250,172390050
Lot NumberA0433944 Certificate TypeCertificate of Analysis Date12/09/2022 Catalog Number172390010,172390250,172390050
compliance-icons
Product Identifier
  • NBD chloride
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : 4-chloro-7-nitro-2,1,3-benzoxadiazole

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
NBD chloride, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.