missing translation for 'onlineSavingsMsg'
Learn More

NBMPR

Product Code. 18025992
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Standard H3 and H4 agonist (H3 > H4)

An extremely potent, high affinity agonist at H3 and H4 receptors (Ki values are 0.3 and 2.7 nM respectively). Induces shape change in eosinophils with an EC50 of 25 nM. Centrally active following systemic administration.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 32385-58-3
Molecular Formula C6H12Br2N4S
Molecular Weight (g/mol) 332.06
MDL Number MFCD00153816
InChI Key DOBOYMKCRRLTRF-UHFFFAOYSA-N
Synonym imetit dihydrobromide, imetit hydrobromide, s-2-imidazol-4-yl ethyl isothiourea, s-2-1h-imidazol-4-yl ethyl isothiourea dihydrobromide, 2-1h-imidazol-4-yl ethyl carbamimidothioate dihydrobromide, s-2-imidazol-4-yl ethyl isothiourea dihydrobromide, imetit dihydrobromide hplc, s-2-imidazol-4-yl ethyl isothiourea 2hbr, s-2-imidazol-4-yl ethyl isothiourea dihydrochloride, 2-2-1h-imidazol-5-yl ethyl isothiourea dihydrobromide
PubChem CID 11957573
ChEBI CHEBI:64151
IUPAC Name {[2-(1H-imidazol-5-yl)ethyl]sulfanyl}methanimidamide dihydrobromide
SMILES Br.Br.NC(=N)SCCC1=CN=CN1

Specifications

Quantity 10 mg
Formula Weight 332.06
Percent Purity >98%
Chemical Name or Material Imetit dihydrobromide
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.