missing translation for 'onlineSavingsMsg'
Learn More

Bochem™ Needle Holder KOLLE with Needle

Manufactured from stainless steel. BOCHEM™ Needle Holder KOLLE with Needle includes needle.

Brand:  Bochem™ 12006

Product Code. 10556110

  • 15.70 € / Each
In Stock
to see stock.
Edge
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Stainless stee
  • Includes needle
Specifications

Specifications

90 mm
Needle holder KOLLE
1 Ea.
Stainless Steel
With needle
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
View accessory products
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Bochem™ Needle Holder KOLLE with Needle > Diameter: 6mm

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.