missing translation for 'onlineSavingsMsg'
Learn More

Nefiracetam, Tocris Bioscience™

Product Code. 18762584
Click to view available options
Quantity:
100 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Cognitive enhancer

Cognitive enhancer that displays various pharmacological effects. Activates L/N-type calcium channels, cholinergic, monoaminergic and GABAergic systems. Displays potent neuroprotective action in the retinal ischemia-reperfusion model in vivo.

Specifications

CAS 77191-36-7
Molecular Formula C14H18N2O2
Quantity 100 mg
Synonym nefiracetam, n-2,6-dimethylphenyl-2-2-oxopyrrolidin-1-yl acetamide, translon, nefiracetam inn, dmppa, motiva, nefiracetamum inn-latin, n-2,6-dimethylphenyl-2-oxo-1-pyrrolidineacetamide, unii-1jk12gx30n, ccris 6729
InChI Key NGHTXZCKLWZPGK-UHFFFAOYSA-N
SMILES CC1=C(C(=CC=C1)C)NC(=O)CN2CCCC2=O
IUPAC Name N-(2,6-dimethylphenyl)-2-(2-oxopyrrolidin-1-yl)acetamide
Molecular Weight (g/mol) 246.31
PubChem CID 71157
Formula Weight 246.3
Percent Purity >99%
Chemical Name or Material Nefiracetam
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.