Learn More
Novus Biologicals™ Negative Control oligodeoxynucleotide Human
Brand:  Novus Biologicals™ NBP2-31135
Product Code. 18113275
  • 237.00 € / 0.10mg
Explore more special offers
Specifications
1 μg/μL
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
0.1 mg
Negative Control Oligodeoxynucleotide