missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NEIL1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 12145-1-AP-150UL
This item is not returnable.
View return policy
Description
NEIL1 belongs to a class of DNA glycosylases homologous to the bacterial Fpg/Nei family. These glycosylases initiate the first step in base excision repair by cleaving bases damaged by reactive oxygen species and introducing a DNA strand break via the associated lyase reaction.Specifications
NEIL1 | |
Polyclonal | |
Unconjugated | |
NEIL1 | |
DNA glycosylase/AP lyase Neil1, Endonuclease 8 like 1, Endonuclease VIII like 1, FPG1, hFPG1, NEH1, Nei homolog 1, Nei like protein 1, NEI1, NEIL1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
367090, 72774, 79661 | |
-20°C | |
Liquid |
Western Blot | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q8K4Q6, Q96FI4 | |
Neil1 | |
NEIL1 Fusion Protein Ag2793 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction