missing translation for 'onlineSavingsMsg'
Learn More

Dr.Weigert™ Neodisher LaboClean A 8

Neodisher LaboClean A8 - Powder Detergent alkaline cleaning is to be used for cleaning and washing glassware. Applications include pathology, parasitology, dairy and food industries.

14.00 € - 119.00 €

Specifications

Product Type Neodisher A8
Form Powder
pH 12 to 12.4
Sterility Non-sterile
Composition Disodium metasilicate pentahydrate; sodium hydroxide; troclosene sodium; Sodium carbonate
View More Specs

Products 2
Product Code Brand Volume (Metric) Price Quantity & Availability  
Product Code Brand Volume (Metric) Price Quantity & Availability  
11717105
View Documents
Dr.Weigert™
410287
1 kg
14.00 €
1kg
Please to purchase this item. Need a web account? Register with us today!
11713733
View Documents
Dr.Weigert™
410276
10 kg
119.00 €
10kg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

  • Alkaline detergent In powder form
  • neodisher LaboClean A 8 is used for cleaning in washer disinfection
  • chlorine-based bleaching agents and phosphates, Free of surfactants
  • suitable for residue-free cleaning of tissue cultures and cell layersDosage of between 2-5 g/L
    Specifications

    Specifications

    Neodisher A8
    12 to 12.4
    Disodium metasilicate pentahydrate; sodium hydroxide; troclosene sodium; Sodium carbonate
    Removes Blood, Serum, Food Residues, Pharmaceuticals, Paints, Varnishes, Gelatin and Other Organic Residues
    Powder
    Non-sterile
    2 to 5 g/L
    Videos
    Safety and Handling

    Safety and Handling

    Product Identifier
    • neodisher LaboClean A 8

    Signal Word
    • Danger

    Hazard Category
    • Substances and mixtures corrosive to metals Category 1
    • Skin corrosion/irritation Category 1 A
    • Serious eye damage/eye irritation Category 1
    • Specific target organ toxicity Category 3
    • LONG-TERM AQUATIC HAZARD Chronic 2

    Hazard Statement
    • H290-May be corrosive to metals.
    • H314-Causes severe skin burns and eye damage.
    • H335-May cause respiratory irritation.
    • H411-Toxic to aquatic life with long lasting effects.
    • EUH031-Contact with acids liberates toxic gas.

    Precautionary Statement
    • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
    • P273-Avoid release to the environment.
    • P280-Wear protective gloves/protective clothing/eye protection/face protection.
    • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
    • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    • P310-Immediately call a POISON CENTER/doctor/.

    Supplemental information
    • MIXTURE LIST-Contains : disodium metasilicate pentahydrate, sodium carbonate, sodiumhydroxide, troclosene sodium

    SDS
    Documents

    Documents

    Product Certifications
    Product Content Correction

    Your input is important to us. Please complete this form to provide feedback related to the content on this product.

    Product Title

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

    Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.