missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dr.Weigert™ Neodisher Z
Neodisher Z - Acidic neutralising agent liquid. Neutralizes by adding to the first rinse after use of an alkaline detergent during automated reprocessing. Free of surfactants and phosphates.
74.40 € - 165.00 €
Specifications
Product Type | Neodisher Z |
---|---|
Form | Liquid |
pH | 3 to 2.6 |
Sterility | Non-sterile |
Composition | Citric acid, 2-octyl-2 H-isothiazol-3-one |
Product Code | Brand | Container Type | Volume (Metric) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Container Type | Volume (Metric) | Price | Quantity & Availability | ||||
10140021
|
Dr.Weigert™
420233 |
Canister (Elongated) | 5 L |
74.40 €
5 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11748393
|
Dr.Weigert™
420235 |
Canister (High Form) | 5 L |
165.00 €
5 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- Based on citric acid
- neodisher Z is used for neutralizing alkaline residues in washer disinfection
- Without phosphates, phosphoric acid, nitrogens compounds and surfactants
- Dosage of between 2-4 mL/L
- Gentle on materials
- Laboratory glassware in medical, industrial and water laboratories
Specifications
Neodisher Z | |
3 to 2.6 | |
Citric acid, 2-octyl-2 H-isothiazol-3-one | |
5 L |
Liquid | |
Non-sterile | |
1 to 5 mL/L |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- neodisher Z
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
Hazard Statement
- H319-Causes serious eye irritation.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
- MIXTURE LIST-Contains : citric acid, anhydrous