missing translation for 'onlineSavingsMsg'
Learn More

Neodymium Boron Iron, REacton™

Product Code. p-4782011
Click to view available options
Quantity:
50 g
250 g
2 x 500 g
Unit Size:
250g
500g
50g
This item is not returnable. View return policy

Product Code. 11396888

Brand: Thermo Scientific Alfa Aesar 013642.18

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Nd:B:Fe; 15:8:77 atomic%

Neodymium Boron Iron applied to manufacture head actuators for computer hard disks, magnetic resonance imaging (MRI), magnetic guitar pickups, loudspeakers and headphones, mechanical e-cigarette firing switches and magnetic bearings and couplings.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Neodymium Boron Iron
Name Note Magnet Alloy Additive
Color Gray-Silver
Physical Form Broken Lump
Quantity 50 g
Molecular Formula Nd:B:Fe; 15:8:77 atomic%
MDL Number MFCD02091729
Synonym Magnet Alloy Additive
Solubility Information Insoluble in water.
Odor Odorless
Appearance Silver gray broken lump
Density 7.5g/cm3
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.