missing translation for 'onlineSavingsMsg'
Learn More

Neodymium(III) chloride, anhydrous, 99.9% (REO), Thermo Scientific Chemicals

84.62 € - 942.69 €

Chemical Identifiers

CAS 10024-93-8
MDL Number MFCD00011132
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11373457
SDS View Documents
Thermo Scientific Alfa Aesar
018680.06
5 g
84.62 €
5g
Please to purchase this item. Need a web account? Register with us today!
11353457
SDS View Documents
Thermo Scientific Alfa Aesar
018680.14
25 g
262.31 €
25g
Please to purchase this item. Need a web account? Register with us today!
11363457
SDS View Documents
Thermo Scientific Alfa Aesar
018680.22
100 g
942.69 €
100g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Neodymium(III) chloride is used as a precursor for the production of neodymium metal. It is used as a catalyst and accelerates polymerization of various dienes such as polybutylene, polybutadiene, and polyisoprene. It has luminescence property and is widely used as fluorescent label in organic molecules, thereby helping in easy tracking of the compound using fluorescence microscope during various physical and chemical reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

10024-93-8
MFCD00011132

Specifications

784°C
Powder
14,6450
250.6
Odorless
4.134 g/mL
Neodymium(III) chloride, Anhydrous
10024-93-8
MFCD00011132
Soluble in water and ethanol.
99.9%
Hygroscopic
(REO)
Videos
Safety and Handling

Safety and Handling

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

missing translation for 'einecsNumber' : 233-031-5

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

Product Identifier
  • Neodymium chloride

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

Supplemental information
  • MIXTURE LIST-Contains : Neodymium chloride (NdCl3)

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.