missing translation for 'onlineSavingsMsg'
Learn More

Neodymium(III) chloride, ultra dry, 99.99% (REO), Thermo Scientific Chemicals

Product Code. 11307867
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Neodymium(III) chloride is used as a precursor for the production of neodymium metal. It is used as a catalyst and accelerates polymerization of various dienes such as polybutylene, polybutadiene, and polyisoprene. It has luminescence property and is widely used as fluorescent label in organic molecules, thereby helping in easy tracking of the compound using fluorescence microscope during various physical and chemical reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Ampouled under argon
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10024-93-8
MDL Number MFCD00011132

Specifications

Melting Point 784°C
Physical Form Powder
Quantity 25 g
Merck Index 14,6450
Solubility Information Soluble in water and ethanol.
Formula Weight 250.6
Percent Purity 99.99%
Odor Odorless
Packaging Ampule
Sensitivity Hygroscopic
Density 4.134 g/mL
Assay (REO)
Chemical Name or Material Neodymium(III) chloride, Ultra dry
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'