missing translation for 'onlineSavingsMsg'
Learn More

Neodymium(III) fluoride, anhydrous, 99.9% (REO), Thermo Scientific Chemicals

147.74 € - 450.24 €

Chemical Identifiers

CAS 13709-42-7
Molecular Formula F3Nd
Molecular Weight (g/mol) 201.24
MDL Number MFCD00011135
InChI Key XRADHEAKQRNYQQ-UHFFFAOYSA-K
IUPAC Name neodymium(3+) trifluoride
SMILES [F-].[F-].[F-].[Nd+3]
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11336308
SDS View Documents
Thermo Scientific Alfa Aesar
011246.14
25 g
147.74 €
25g
Please to purchase this item. Need a web account? Register with us today!
11326308
SDS View Documents
Thermo Scientific Alfa Aesar
011246.22
100 g
450.24 €
100g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Neodymium(III) fluoride, is used extensively in the preparation and study of fluorinated glasses.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

13709-42-7
201.24
XRADHEAKQRNYQQ-UHFFFAOYSA-K
[F-].[F-].[F-].[Nd+3]
F3Nd
MFCD00011135
neodymium(3+) trifluoride

Specifications

1410°C
Purple
F3Nd
MFCD00011135
XRADHEAKQRNYQQ-UHFFFAOYSA-K
neodymium(3+) trifluoride
201.24
Odorless
6.5 g/mL
Neodymium(III) fluoride, Anhydrous
13709-42-7
Crystalline Lump
NdF3
Soluble in aqueous solutions.
[F-].[F-].[F-].[Nd+3]
201.24
99.9%
Hygroscopic
(REO)
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

H314

missing translation for 'dotInformation' : Hazard Class: 8; Packaging Group: III

missing translation for 'einecsNumber' : 237-253-3

missing translation for 'rtecsNumber' : QO9500000

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.