missing translation for 'onlineSavingsMsg'
Learn More

Neodymium(III) nitrate hydrate, REacton™, 99.99% (REO), Thermo Scientific Chemicals

44.32 € - 320.27 €

Chemical Identifiers

CAS 13746-96-8
Molecular Formula N3NdO9
Molecular Weight (g/mol) 330.25
MDL Number MFCD00149798
InChI Key CFYGEIAZMVFFDE-UHFFFAOYSA-N
Synonym neodymium iii nitrate hydrate, neodymium nitrate hydrate, acmc-20ak53, 3no3.nd.h2o, neodymium nitrate-water 1/3/1, neodymium 3+ ion hydrate trinitrate, neodymium 3+ hydrate trinitrate, neodymium iii nitrate hydrate, reacton, nitric acid,neodymium 3+ salt, hydrate 8ci,9ci
PubChem CID 71311426
SMILES [Nd+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11316308
SDS View Documents
Thermo Scientific Alfa Aesar
011245.06
5 g
44.32 €
5g
Please to purchase this item. Need a web account? Register with us today!
11396298
SDS View Documents
Thermo Scientific Alfa Aesar
011245.14
25 g
159.80 €
25g
Please to purchase this item. Need a web account? Register with us today!
11306308
SDS View Documents
Thermo Scientific Alfa Aesar
011245.22
100 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

It is used as pharmaceutical intermediates. It is used in the selective PVC membrane sensors. The sensor with ionophore (L1) exhibits significantly enhanced selectivity towards neodymium (III).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

13746-96-8
330.25
CFYGEIAZMVFFDE-UHFFFAOYSA-N
71311426
N3NdO9
MFCD00149798
neodymium iii nitrate hydrate, neodymium nitrate hydrate, acmc-20ak53, 3no3.nd.h2o, neodymium nitrate-water 1/3/1, neodymium 3+ ion hydrate trinitrate, neodymium 3+ hydrate trinitrate, neodymium iii nitrate hydrate, reacton, nitric acid,neodymium 3+ salt, hydrate 8ci,9ci
[Nd+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O

Specifications

∽40°C
Blue
99.99% (REO)
MFCD00149798
Soluble in water.
[Nd+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O
330.25
330.25 (Anhydrous)
Hygroscopic
Neodymium(III) nitrate hydrate
13746-96-8
Wet Crystalline Aggregates
N3NdO9
neodymium iii nitrate hydrate, neodymium nitrate hydrate, acmc-20ak53, 3no3.nd.h2o, neodymium nitrate-water 1/3/1, neodymium 3+ ion hydrate trinitrate, neodymium 3+ hydrate trinitrate, neodymium iii nitrate hydrate, reacton, nitric acid,neodymium 3+ salt, hydrate 8ci,9ci
CFYGEIAZMVFFDE-UHFFFAOYSA-N
neodymium(3+);trinitrate;hydrate
71311426
Odorless
Violet crystals
Videos
Safety and Handling

Safety and Handling

P210-P220-P221-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c

H272-H315-H319-H335

missing translation for 'dotInformation' : DOT Class: 5.1, Packaging Group: III

missing translation for 'einecsNumber' : 233-153-9

missing translation for 'rtecsNumber' : OJ6125000

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.