missing translation for 'onlineSavingsMsg'
Learn More
Neodymium(III) oxide, 99.9% (REO), Thermo Scientific Chemicals Product Code.: 11393338

Neodymium(III) oxide, 99.9% (REO), Thermo Scientific Chemicals

Product Code. 11393338
100 g
Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy

Product Code. 11393338

Brand: Thermo Scientific Alfa Aesar 036275.22

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Neodymium is used in coloring glasses and for doping some glass lasers since it transmits 90% of the blue, green, and red light rays. Neodymium compounds are used in glass coloring, permanent magnets, capacitors, infrared filters, resonators, Catalysis (diene polymerization), laser materials. Neodymium(III) oxide is used to dope glass, including sunglasses, to make solid-state lasers, and to color glasses and enamels.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Identificatori chimici

CAS 1313-97-9
Molecular Formula Nd2O3
Molecular Weight (g/mol) 336.48
MDL Number MFCD00011134
InChI Key PLDDOISOJJCEMH-UHFFFAOYSA-N
IUPAC Name dineodymium(3+) trioxidandiide
SMILES [O--].[O--].[O--].[Nd+3].[Nd+3]

Specifica

Melting Point 2272°C
Color Blue
Physical Form Sintered Lump
Quantity 100 g
Assay Percent Range 99.9%
Solubility Information Insoluble in water. Soluble in mineral acids.
Formula Weight 336.48
Appearance Light blue 3-12mm Sintered Lump
Density 7.24
Chemical Name or Material Neodymium(III) oxide

RUO – Research Use Only

Correzione del contenuto del prodotto

Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.

Titolo del prodotto
Neodymium(III) oxide, 99.9% (REO), Thermo Scientific Chemicals >

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.

Grazie per averci aiutato a migliorare il nostro sito web. Il vostro feedback è stato inviato