Learn More
Neodymium(III) oxide, 99.9% (REO), Thermo Scientific Chemicals
128.00 € - 460.29 €
Chemical Identifiers
CAS | 1313-97-9 |
---|---|
Molecular Formula | Nd2O3 |
Molecular Weight (g/mol) | 336.48 |
MDL Number | MFCD00011134 |
InChI Key | PLDDOISOJJCEMH-UHFFFAOYSA-N |
IUPAC Name | dineodymium(3+) trioxidandiide |
SMILES | [O--].[O--].[O--].[Nd+3].[Nd+3] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11303348
|
Thermo Scientific Alfa Aesar
036275.14 |
25 g | N/A | ||||||
11393338
|
Thermo Scientific Alfa Aesar
036275.22 |
100 g |
460.29 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Neodymium is used in coloring glasses and for doping some glass lasers since it transmits 90% of the blue, green, and red light rays. Neodymium compounds are used in glass coloring, permanent magnets, capacitors, infrared filters, resonators, Catalysis (diene polymerization), laser materials. Neodymium(III) oxide is used to dope glass, including sunglasses, to make solid-state lasers, and to color glasses and enamels.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
1313-97-9 | |
336.48 | |
PLDDOISOJJCEMH-UHFFFAOYSA-N | |
[O--].[O--].[O--].[Nd+3].[Nd+3] |
Nd2O3 | |
MFCD00011134 | |
dineodymium(3+) trioxidandiide |
Specifications
2272°C | |
Blue | |
99.9% | |
MFCD00011134 | |
PLDDOISOJJCEMH-UHFFFAOYSA-N | |
dineodymium(3+) trioxidandiide | |
336.48 | |
7.24 g/cm3 at 20°C |
1313-97-9 | |
Sintered Lump | |
Nd2O3 | |
Insoluble in water. Soluble in mineral acids. | |
[O--].[O--].[O--].[Nd+3].[Nd+3] | |
336.48 | |
Light blue 3-12mm Sintered Lump | |
Neodymium(III) oxide |
Safety and Handling
missing translation for 'einecsNumber' : 215-214-1
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.