missing translation for 'onlineSavingsMsg'
Learn More
Neodymium(III) oxide, REacton™, 99.9% (REO), Thermo Scientific Chemicals Product Code.: 11366308

Neodymium(III) oxide, REacton™, 99.9% (REO), Thermo Scientific Chemicals

Product Code. 11366308
250 g
Click to view available options
Quantity:
50 g
250 g
1 kg
This item is not returnable. View return policy

Product Code. 11366308

Brand: Thermo Scientific Alfa Aesar 011248.30

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Neodymium(III) oxide is used as a catalyst in polymer chemistry. Glasses doped with neodymium(III) oxide are dichroic in nature and is used in welding goggles and other colored glasses. It finds use in solid-state lasers, carbon arc-light electrodes and enamels. It serves as the starting material for the synthesis of other neodymium(III) compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1313-97-9
Molecular Formula Nd2O3
Molecular Weight (g/mol) 336.48
MDL Number MFCD00011134
InChI Key PLDDOISOJJCEMH-UHFFFAOYSA-N
IUPAC Name dineodymium(3+) trioxidandiide
SMILES [O--].[O--].[O--].[Nd+3].[Nd+3]

Specifications

Melting Point 2272°C
Color Yellow
Physical Form Powder
Quantity 250 g
Assay Percent Range 99.9% (REO)
Solubility Information Insoluble in water. Soluble in acids.
Formula Weight 336.48
Appearance Light blue powder
Density 7.24 g/cm3 at 20°C
Chemical Name or Material Neodymium(III) oxide

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Neodymium(III) oxide, REacton™, 99.9% (REO), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.