Learn More
Neodymium(III) oxide, REacton™, 99.999% (REO), Thermo Scientific Chemicals
Description
Neodymium(III) oxide is used as a catalyst in polymer chemistry. Glasses doped with neodymium(III) oxide are dichroic in nature and is used in welding goggles and other colored glasses. It finds use in solid-state lasers, carbon arc-light electrodes and enamels. It serves as the starting material for the synthesis of other neodymium(III) compounds.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Melting Point | 2272°C |
Color | Blue |
Physical Form | Powder |
Quantity | 25 g |
Merck Index | 14,6450 |
Solubility Information | Insoluble in water. Soluble in acids. |
Formula Weight | 336.48 |
Percent Purity | 99.999% |
Grade | Reacton™ |
Density | 7.24 g/mL |
Show More |
Product Suggestions
Customers who viewed this item also viewed.
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
M20I015 | Certificate of Analysis | 12/04/2022 | 045566.14,045566.22 |
R26B014 | Certificate of Analysis | 07/12/2021 | 045566.14,045566.22 |
N17G108 | Certificate of Analysis | 07/12/2021 | 045566.14,045566.22 |
Z07C020 | Certificate of Analysis | 29/11/2021 | 045566.14,045566.22 |
X25C028 | Certificate of Analysis | 29/11/2021 | 045566.14,045566.22 |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.