missing translation for 'onlineSavingsMsg'
Learn More
Neodymium(III) oxide, REacton™, 99% (REO), Thermo Scientific Chemicals Product Code.: 11373768

Neodymium(III) oxide, REacton™, 99% (REO), Thermo Scientific Chemicals

Product Code. 11373768
100 g
Click to view available options
Quantity:
25 g
100 g
500 g
This item is not returnable. View return policy

Product Code. 11373768

Brand: Thermo Scientific Alfa Aesar 040252.22

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

ceramic capacitors, coloring glass, carbon arc-light electrodes, color TV tubes, dehydrogenation catalyst.Neodymium(III) oxide is an inorganic compound containing inner transition metal neodymium from lanthanides series in +3 oxidation state and oxide in 2:3 ratio. It adopts a hexagonal crystal structure. It is also known as neodymium oxide, neodymiumtrioxide, neodymium(3+)oxide, neodymium sesquioxide and dineodymiumtrioxide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Ceramic capacitors, coloring glass, carbon arc-light electrodes, color TV tubes, dehydrogenation catalyst
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1313-97-9
Molecular Formula Nd2O3
Molecular Weight (g/mol) 336.48
MDL Number MFCD00011134
InChI Key PLDDOISOJJCEMH-UHFFFAOYSA-N
IUPAC Name dineodymium(3+) trioxidandiide
SMILES [O--].[O--].[O--].[Nd+3].[Nd+3]

Specifications

Melting Point 2272°C
Physical Form Powder
Quantity 100 g
Merck Index 14,6450
Solubility Information Insoluble in water; Soluble in acids.
Formula Weight 336.48
Percent Purity 99%
Grade Reacton™
Density 7.24 g/mL
Assay (REO)
Chemical Name or Material Neodymium(III) oxide
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Neodymium(III) oxide, REacton™, 99% (REO), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.